Imprimer
Pilons pour microtubes de 0,5 et 1,5 ml, Dstroy®-Sticks
Fournisseur: BIOZYM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
710388 710389 710398 710399
BZYM710388EA 0 EUR
BZYM710388 BZYM710389 BZYM710398 BZYM710399
Pilons pour microtubes de 0,5 et 1,5 ml, Dstroy®-Sticks
Mortiers et pilons Pilons
These Dstroy®-Sticks and associated reaction tubes are made of polypropylene.
  • Free from RNases, DNases, DNA and pyrogens
Mostly used in the applications homogenising small amounts of cell materials in a reaction vessel and resuspending protein or DNA precipitates in appropriate buffer solutions.