Imprimer
HPLC columns, Acclaim™ PepMap™ C18
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
160434 164199 164212 164213 164534 164535 164537 164560 164568 164569 164570 164572
555-6891EA 700 EUR
555-6891 555-6797 555-6800 555-6801 555-6815 555-6783 555-6765 555-6759 555-6758 555-6767 555-6768 555-6770
HPLC columns, Acclaim™ PepMap™ C18
Colonnes pour chromatographie
Acclaim™ PepMap™ C18 columns are ideal for high-resolution analysis of tryptic, natural, and synthetic peptides. The columns are often applied for LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology. Due to their high loading capacity, the columns are suitable for the analysis of low abundant peptides in complex proteomics samples.
  • High resolution protein identification for biomarker discovery
  • Highest sensitivity in LC-MS