Imprimer
Seringues HPLC
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
365ILT91 365GL251 365JL281 365JLT61 365ILT21
555-8225EA 247 EUR
555-8225 555-8443 555-8236 555-8239 555-8224
Seringues HPLC
Seringues Seringues de chromatographie
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
  • Easy and accurate
  • Reproducible manual injections
Applications include industrial chromatography.