Imprimer
LiChrospher®, HPLC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LISP60-5-150AEA 845 EUR
LISP60-5-150A LISP60-5-250A LISPCN-5-250A LISPOH-5-250A LISP100-5-150A LISP100-5-250A LISP60-5-125AF HICHLISP100-5100AF HICHLISP100-5100AM HICHLISP100-5100AS LISP100-5-10C5 LISPOH-5-100AF LISPOH-5-10C5 LISPOH-5-10CM5 LISPOH-5-125AF LISPOH-5-150AM LISPOH-5-250AF LISPOH-5-250AM HICHLISP60-5-10CP3 LISP60-5-250P LISPOH510C5SK LISPOH-5-10CE5 LISPOH-5-250P
LiChrospher®, HPLC Columns
Colonnes pour chromatographie
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns