Imprimer
HPLC columns, HyPURITY™ ADVANCE
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21005-254630 21005-154630 21005-054630 21005-102130 21005-104630
555-7032EA 0 EUR
555-7032 555-7029 555-7023 555-7024 555-7026
HPLC columns, HyPURITY™ ADVANCE
Colonnes pour chromatographie
This reversed phase column provides a polar embedded stationary phase for alternate selectivity. These groups block surface silanols from interacting with polar analytes and offers improved chromatographic performance with broad applicability and improved peak shape.
  • Ideal for high throughput fast analyses
  • Enhanced aqueous stability
  • Exceptional peak shape for acids, bases and chelators