Imprimer
Workers jackets, C.I., Design C, black
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
849940210M 849940210S 849940210XL 849940210XS 8499402312XL 849940231L 849940231M 849940231S 849940231XL 849940231XS 8499403312XL 849940331L 849940331M 849940331S 849940331XL 849940331XS 8499404402XL 849940440L 849940440M 849940440S 849940440XL 849940440XS 8499405202XL 849940520L 849940520M 849940520S 849940520XL 849940520XS 8499405302XL 849940530L 849940530M 849940530S 849940530XL 849940530XS 8499405392XL 849940539L 849940539M 849940539S 849940539XL 849940539XS 8499406402XL 849940640L 8499401312XL 849940131L 849940131M 849940131S 849940131XL 849940131XS 8499402102XL 849940210L 849940640M 849940640S 849940640XL 849940640XS 8499407302XL 849940730L 849940730M 849940730S 849940730XL 849940730XS 8499409402XL 849940940L 849940940M 849940940S 849940940XL 849940940XS 8499409412XL 849940941L 849940941M 849940941S 849940941XL 849940941XS
KWIN849940210MEA 0 EUR
KWIN849940210M KWIN849940210S KWIN849940210XL KWIN849940210XS KWIN8499402312XL KWIN849940231L KWIN849940231M KWIN849940231S KWIN849940231XL KWIN849940231XS KWIN8499403312XL KWIN849940331L KWIN849940331M KWIN849940331S KWIN849940331XL KWIN849940331XS KWIN8499404402XL KWIN849940440L KWIN849940440M KWIN849940440S KWIN849940440XL KWIN849940440XS KWIN8499405202XL KWIN849940520L KWIN849940520M KWIN849940520S KWIN849940520XL KWIN849940520XS KWIN8499405302XL KWIN849940530L KWIN849940530M KWIN849940530S KWIN849940530XL KWIN849940530XS KWIN8499405392XL KWIN849940539L KWIN849940539M KWIN849940539S KWIN849940539XL KWIN849940539XS KWIN8499406402XL KWIN849940640L KWIN8499401312XL KWIN849940131L KWIN849940131M KWIN849940131S KWIN849940131XL KWIN849940131XS KWIN8499402102XL KWIN849940210L KWIN849940640M KWIN849940640S KWIN849940640XL KWIN849940640XS KWIN8499407302XL KWIN849940730L KWIN849940730M KWIN849940730S KWIN849940730XL KWIN849940730XS KWIN8499409402XL KWIN849940940L KWIN849940940M KWIN849940940S KWIN849940940XL KWIN849940940XS KWIN8499409412XL KWIN849940941L KWIN849940941M KWIN849940941S KWIN849940941XL KWIN849940941XS
Workers jackets, C.I., Design C, black
Tenue de travail
Industrial jacket made of 65% polyester and 35% cotton, with soft nap on reverse side. The black jacket is available with different accent and piping colours.
  • Zip closure
  • Two chest pockets, one double with flap and velcro closure
  • Two front pockets, glove pocket, sleeve pocket, concealed ID-card pocket
  • Mobile phone pocket with loop for earpiece, concealed pen pocket, two inside pockets
  • Adjustable waist and cuffs, rounded elbows
  • Choice of 12 profille colours and 13 piping colours
Weight: 260 g/m²