Imprimer
Avantor® Hichrom, Chiral Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CHI-DL-PGC-10C5EA 0 EUR
CHI-DL-PGC-10C5 CHI-DL-PGC-10CP3 HICHCHI-DLPGC250AS HICHCHI-DLPGC250SP CHI-D-PGC-10CP3 HICHCHI-D-PGC250AS CHI-D-PGC-250SP CHI-L-LEUC-10C5 CHI-L-LEUC-10CP3 CHI-L-LEUC-250A HICHCHI-LLEUC250AS HICHCHI-LLEUC250SP CHI-L-PGC-10CP3 HICHCHI-L-PGC250AS CHI-L-PGC-250SP
Avantor® Hichrom, Chiral Columns
Colonnes pour chromatographie
Chiral columns are designed for the analysis of a wide range of enantiomers of pharmaceutical and agrochemical origin. This column contains a chiral dinitrophenyltartramide moiety is bonded to the silica surface through a propyl spacer group.
  • Chiral Pirkle-type
  • High efficiency and unique covalent bonding
  • Enantiomer order elution inversion
Substitution of L-phenylglycine for D-phenylglycine, the order of elution of the chiral peaks can be reversed. This can be useful in assigning peak identity or ensuring prior elution of the minor enantiomer, thus allowing its more accurate determination.