Imprimer
Plaque pour PCR 96 puits, profil abaissé, demi-jupe, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PCR-96-LP-AB-C PCR-96-LP-AB-W PCR-96LPABCPBC
732-1015EA 208 EUR
732-1015 732-1017 AXYGPCR-96LPABCPBC
Plaque pour PCR 96 puits, profil abaissé, demi-jupe, Axygen®
Microplaques Plaques PCR
Axygen® 96 well, PP, low profile PCR microplates are designed to fit ABI Real-Time PCR thermal cyclers and sequencers.
  • Optimised for Fast PCR: Low profile design results in faster and more efficient PCR reactions
  • Reduced volume minimises potential evaporation
  • Ultra-thin and consistent wall thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified RNase, DNase and endotoxin-safe for highest sample integrity
  • Can be sealed with 8/12 strip caps, silicone AxyMat™ or sealing film
  • AxyMat™ sealing mats autoclavable and reusable
Compatible with (in addition to standard thermal cycling): ABI Fast PCR 7500, 7900, 9800, Veriti and StepOne.