Imprimer
Xylene substitute, Neo-Clear®, Sigma-Aldrich®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.09843.5000 1.09843.9025
1.09843.5000EA 125 EUR
1.09843.5000 1.09843.9025
Xylene substitute, Neo-Clear®, Sigma-Aldrich®
Colorants Réactifs pour la microscopie et la coloration
Neo-Clear® est une alternative non dangereuse au xylène. Il est pratiquement inodore, compatible avec la plupart des solvants haute pureté et se caractérise par des taux d'évaporation plus faibles que le xylène, ce qui en fait un produit moins dangereux pour la santé.
Neo-Clear® is an aliphatic hydrocarbon mixture that can substitute the same applications as xylene in the histo-medical lab - histoprocessing, deparaffination and clearing after dehydration in staining process
It is not necessary to change methods or incubation times when using Neo-Clear® instead of xylene. As the evaporation rate of Neo-Clear® is lower than xylene, it has less odour and the disturbing smell in the lab is reduced significantly. Neo-Clear® is more sensitive against water in solvents (turbidity can occur), so, it is essential to avoid technical qualities of solvents. It is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes.