Imprimer
Shirts, long sleeves, GEN-Y Cocona®
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1009669402XL 1009669403XL 100966940L 100966940M 100966940S 100966940XL 100966940XS
KWIN1009669402XLEA 0 EUR
KWIN1009669402XL KWIN1009669403XL KWIN100966940L KWIN100966940M KWIN100966940S KWIN100966940XL KWIN100966940XS
Shirts, long sleeves, GEN-Y Cocona®
Tenue de travail
Long sleeved shirts made of 44,5% polyester (Cocona®) and 55,5% standard polyester. Cocona® fabric transports moisture, absorbs odour, dries very quickly and offers UV protection.
  • Flatlock stitching detail
  • Drop shoulder at front
  • V-stitch at neck, taped neck edge
  • Y embroidery on shoulder
Weight: 145 g/m²