Imprimer
Underwear set, 7416 UE
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1010319402XL 101031940L 101031940S 101031940XL 101031940XS
KWIN1010319402XLEA 33.9 EUR
KWIN1010319402XL KWIN101031940L KWIN101031940S KWIN101031940XL KWIN101031940XS
Underwear set, 7416 UE
Tenue de travail
This set consists of a shirt with long sleeves and a pair of long johns. Both are made of 100% polyester.
  • Shirt with round neck
  • Flatlock stitching details in contrasting colour
Weight: 178 g/m²