Imprimer
Face shields, chin guard system, Connect®
Fournisseur: CENTURION SAFETY PRODUCTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S910 S911 S912
CENUS910EA 22.3 EUR
CENUS910 111-1580 CENUS912
Face shields, chin guard system, Connect®
Écrans de protection
These clear face shields are available with acetate or PC screens.
  • Chinguard screens have holes at bottom in order to attach chinguard
  • Can be used in combination with a browguard when a safety helmet is not required
Certifications: EN 166 B