Imprimer
Rapid antibody labelling kits, VivoBrite™
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
92160 92161 92162
92160.EA 545 EUR
92160. 92161. 92162.
Rapid antibody labelling kits, VivoBrite™
Colorants Colorants d'imagerie in vivo
These antibody labelling kits include amine-reactive near-IR CF™ dye as well as all components required for antibody labelling and purification.
Suitable for small animal in vivo imaging.
Informations de commande: Each kit is sufficient for three labellings.
Attention: This item is temperature sensitive and has specific storage requirements.