Imprimer
Safety Bottle Container
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9.20078.0001 1.40140.0001
9.20078.0001EA 86.4 EUR
9.20078.0001 1.40140.0001
Safety Bottle Container
Portoirs Portoirs pour flacons
These bucket-style bottle carriers are essential safety equipment for transportation of hazardous chemicals.
  • Versions for 2,5 L and 4 L glass bottles
  • Additional bottle protection by a special developed carrier with foam inlay
  • Solvent-resistant materials
  • Leak-proof top cover
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable