Imprimer
Reusable half masks, X-plore® 2100
Fournisseur: Dräger
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
X-plore®
DRAER55881EA 58.4 EUR
DRAER55881
Reusable half masks, X-plore® 2100
Respirateurs Masques réutilisables Demi-masques
The Dräger X-plore® 2100 is the reusable alternative to disposable dust masks for people working frequently in dusty environments. The ergonomic non-slip head harness ensures a good fit, while the elastic straps with an integrated neck hook that are easy to open and close, and allow precise adjustment of the mask. The mask can also be removed without the need to take off the safety helmet.
  • Excellent fit with less leakage rates
  • Low weight
  • Used in various industries as well as in agriculture
Available in two high quality materials such as EPDM or silicone.