Imprimer
Diffusion tubes, with direct indication, for TWA measurements
Fournisseur: Dräger
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8101161 8101301 8101111
DRAE8101161EA 176 EUR
DRAE8101161 DRAE8101301 DRAE8101111
Diffusion tubes, with direct indication, for TWA measurements
Systèmes de détection de gaz
Long-term measurements with diffusion tubes provide integrated measurements that represent the average concentration during the sampling period.
  • Measurements performed between one and eight H
  • Tubes can be used as personal, or area monitors
  • Determine the weight average concentration