Imprimer
Tubing sleeves, FEP, Upchurch Scientific®
Fournisseur: UPCHURCH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-374 F-376 F-370 F-375 F-372
UPCHF-374EA 11.8 EUR
UPCHF-374 UPCHF-376 UPCHF-370 UPCHF-375 UPCHF-372
Tubing sleeves, FEP, Upchurch Scientific®
Tuyaux Tubulures à chromatographie
FEP, natural or coloured.
FEP tubing sleeves are an excellent choice for lower pressure applications. These 1,6" long sleeves facilitate connecting capillary tubing into ports designed for 1/32" Ext.Ø. tubing. A wide selection of appropriate sleeve for capillary Ext.Ø. size. These sleeves can be used at up to 50 °C and have a pressure rating of 1750 psi (121 bar).