Imprimer
Face shields, ArcPro™ anti electric arc systems
Fournisseur: CENTURION SAFETY PRODUCTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S25WCONTAEA S25WCEAEA S506S57 S512S57 S910AEA S25WR820
CENUS25WCONTAEAEA 108 EUR
CENUS25WCONTAEA CENUS25WCEAEA CENUS506S57 CENUS512S57 CENUS910AEA CENUS25WR820
Face shields, ArcPro™ anti electric arc systems
Face Shields
These systems have been designed to protect against arc flash risks posed by electrical faults.
  • Clear PC screen protects against short circuit arcs
  • Green PC screens also feature anti-fog coating
  • Face screens supplied with chin guard made of fibre reinforced nylon
The face screens are available without or in complete kits with helmet.
Certifications: EN 166 1 B 8 9 (EN 170 2-1,2); Arc exposure test according to: GS-ET-29:2010, Class 2 (423 kJ/m²)