Imprimer
HPLC columns, Hypersil™ ODS-2 C18
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
31605-014001 31605-254630 31605-154630 31605-254030 31605-104630 31605-054630 31603-154630 31603-104630 31603-054630 31603-014001
555-5244EA 462 EUR
555-5244 555-5281 555-5228 555-5282 555-5235 555-5239 555-5248 555-5313 555-5317 555-5321
HPLC columns, Hypersil™ ODS-2 C18
Colonnes pour chromatographie
Hypersil™ ODS-2 C18 columns are well established and referenced in HPLC methods. Hypersil™ stationary phases are recognised as an industry standard in HPLC.
  • Reproducibility and high selectivity over wide range of applications
  • Dependable L1 and well tested