Imprimer
Blouson, crafts
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1006295502XL 100629550L 100629550M 100629550S 100629550XL 100629550XS
KWIN1006295502XLEA 56 EUR
KWIN1006295502XL KWIN100629550L KWIN100629550M KWIN100629550S KWIN100629550XL KWIN100629550XS
Blouson, crafts
Tenue de travail
Blouson for handcraft applications. Made of 100% cotton.
  • Chest pocket
  • Band around neck
  • Cuffs with button
  • Belts tie around waist
Weight: 220 g/m²