Imprimer
Single channel pipettes, electronic, variable volume, E4™ XLS™
Fournisseur: RAININ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17012312 17012313 17012314 17012353 17012354 17014486 17014483 17014485 17014491 17014490 17014494 17014484 17014487 17014482 17014488 17014493 17014495 17014489 17014492
RAIN17012312EA 1540 EUR
RAIN17012312 RAIN17012313 RAIN17012314 RAIN17012353 RAIN17012354 RAIN17014486 RAIN17014483 RAIN17014485 RAIN17014491 RAIN17014490 RAIN17014494 RAIN17014484 RAIN17014487 RAIN17014482 RAIN17014488 RAIN17014493 RAIN17014495 RAIN17014489 RAIN17014492
Single channel pipettes, electronic, variable volume, E4™ XLS™
Pipettes Pipetteurs
The E4™ XLS™ is an electronic pipette that can be upgraded and expanded right from the PC. Available as standard versions or with LTS LiteTouch System for lower ejection forces.
  • Large screen, joystick control and carousel menus speed access to all modes and options
  • True Manual™ pipetting mode permits continuous Real Time control of pipetting speed and direction, combined with precise volume measurement
  • Independent control of aspirate, dispense and mix speeds for challenging liquids
  • Blowout and automatic dispensing options for precision pipetting protocols
  • Ergonomic fingerhook and joystick provide maximum comfort
Standard functions include basic pipetting, multi-dispensing, and mixing. Advanced functions include sequential volume programming, fixed volume, titration, dilution and reverse mode pipetting.
Informations de livraison: Supplied with Li-Ion battery and wall power supply. An optional charging stand is available separately.