Imprimer
Industrial gloves, Monsun® 105
Fournisseur: KCL by Honeywell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
105.7 105.8 105.9 105.10
112-1225EA 12.2 EUR
112-1225 112-1226 112-1227 112-1228
Industrial gloves, Monsun® 105
Gants
These tight-fitting knitted cotton liners with palm nitrile coating and knit wrist are ideal for precision jobs.
  • Moisture-repellent, very breathable
  • Glove surface free from silicone (lacquer indifference, test method automotive industry)
  • Particularly close-fitting for applications where sensitivity is needed
  • Allows longer working without showing signs of fatigue
  • Additional wrist protection
Suitable applications include assembly and transportation works, manufacturing of small parts, warehouse and packaging applications, metal processing, precision engineering and electronical as well as automotive industries.
Thickness: 0,75 ± 0,05 mm
Colour: Yellow/white
EN 388 (2111), Cat. II Food contact approved
Certifications: EN 388:2016 (2111X), Cat. II
Food contact approved