Imprimer
Tubing sleeves, PEEK, MicroTight®, Upchurch Scientific®
Fournisseur: UPCHURCH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-180 F-181 F-182 F-183 F-184 F-185 F-185X F-186 F-187 1328 1356
UPCHF-180EA 0 EUR
UPCHF-180 UPCHF-181 UPCHF-182 UPCHF-183 UPCHF-184 UPCHF-185 UPCHF-185X UPCHF-186 UPCHF-187 UPCH1328 UPCH1356
Tubing sleeves, PEEK, MicroTight®, Upchurch Scientific®
Tuyaux Tubulures à chromatographie
PEEK.
PEEK microtight connector kit offers convenience and a cost effective option. These sleeves assist in creating a concentric connection with minimal dead volume. This kit offers a 10 Pack of each tubing sleeve F-180 through F-187, (2) P-770 MicroTight Adapters and (2) MicroTight P-720 Unions.