Imprimer
DPD chlorine testing systems, SwifTest™
Marques : Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2802300 2105660 2802400 2105560
HACH2802300EA 183 EUR
HACH2802300 HACH2105660 HACH2802400 HACH2105560
DPD chlorine testing systems, SwifTest™
Kits de tests environnementaux
  • Fast, easy reagent addition - ideal for high volume testing
  • Just squeeze to add powder, no need to open reagent containers
Informations de livraison: Dispenser is supplied with 1 refill vial with enough reagent for approximately 250 tests.