Imprimer
DrySyn® single- and multi-position cooling blocks, DrySyn COOL and Multi COOL
Fournisseur: ASYNT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ADS10 ADS8
442-0870EA 4260 EUR
442-0870 442-0861
DrySyn® single- and multi-position cooling blocks, DrySyn COOL and Multi COOL
Block Heaters Block Heater/Coolers
For controlled cooling of 25 to 1000 ml flasks without using jacketed reaction vessels. Units allow active temperature control from +150 to −30 °C (depending on circulator specification).
  • Compatible with all standard laboratory circulator or chiller systems and any standard magnetic stirrer or overhead stirrer
  • Compatible with standard laboratory round bottom flasks
  • Precise temperature control of cooling block or flask contents (depending on circulator used)
  • Unattended operation allows for overnight cooled reactions
  • Possible to programme and control cooling/heating ramp rates (depending on circulator used)