Imprimer
Distillation columns, Vigreux, with removable glass jacket
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05133504 05133201 05133304 05133503 05133301
LENZ05133504EA 222 EUR
LENZ05133504 201-1665 201-1666 LENZ05133503 LENZ05133301
Distillation columns, Vigreux, with removable glass jacket
Distillation Components Distillation Columns
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket and NS cone, with removable glass jacket, horizontal and lateral indentations.