Imprimer
Power supply adapters
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
18-1129-58 18-1129-59
18-1129-58EA 126 EUR
18-1129-58 18-1129-59
Power supply adapters
Alimentations électriques
Choice of adapters for connecting 4 or 2 mm leads to 2 or 4 mm output jacks, respectively.
  • Approved for 500 V relative protection ground
  • Adapter set 2 mm M/4 mm F for connecting sheathed 2 mm male plug instrument leads to 4 mm sheathed output jacks
  • Adapter set 4 mm M/2 mm F for connecting sheathed 4 mm male plug instrument leads to 2 mm high voltage output jacks