Imprimer
Colonnes capillaires, TraceGOLD™ TG-SQC
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26070-1420 26070-1300
555-8772EA 978 EUR
555-8772 555-8773
Colonnes capillaires, TraceGOLD™ TG-SQC
Colonnes pour chromatographie
Optimised for system qualification tests for new GC-MS installations or during service/maintenance of an existing instrument.
  • Recommended for benchmark testing only