Imprimer
High quality prismatic optical bench
Fournisseur: DIDALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
POD060100 POD060110 POD060056 POD060090 POD060080
DIDAPOD060100EA 0 EUR
DIDAPOD060100 DIDAPOD060110 DIDAPOD060056 DIDAPOD060090 DIDAPOD060080
High quality prismatic optical bench
Matériel éducatif Matériel éducatif de physique Activités d'optique
Triangular, cast aluminium optical bench
  • Lightweight for easy transport and manoeuvrability
  • Graduated measure to accurately determine distance between components for reproducibility of results
  • Compatible with most triangular riders
  • Adjustable feet for stability