Imprimer
Colonnes capillaires, TracePLOT™ TG-BOND Q+
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26005-6000 26005-6030 26005-6060 26005-6090
555-8867EA 812 EUR
555-8867 555-8866 555-8865 555-8864
Colonnes capillaires, TracePLOT™ TG-BOND Q+
Colonnes pour chromatographie
Intermediate polarity columns for baseline separation of ethane, ethylene and acetylene.
  • Porous divinyl benzene homopolymer
  • Particles incorporated into the walls of the tubing for essentially no particle release