Imprimer
Capillary columns, TraceGOLD™ TG-5MS AMINE
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26097-1300 26097-1420 26097-2230 26097-2960 26097-2970 26097-2980 26097-3960 26097-2840
555-8643EA 642 EUR
555-8643 555-8642 555-8640 555-8698 555-8697 555-8696 555-8694 555-8639
Capillary columns, TraceGOLD™ TG-5MS AMINE
Colonnes pour chromatographie
TraceGOLD™ TG-5MS AMINE GC columns are ideal for the analysis of ppm levels of amines without column priming.
  • Low polarity phase, base optimised
  • Tubing surface is chemically altered to reduce tailing of active basic compounds
  • Allows analysis of neutral or weakly acidic compounds
  • Low bleed at maximum operating temperature