Imprimer
Barrier tips, low retention, ART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2069-05-HR 2139-05-HR 2149E-05-HR 2149P-05-HR 2179-05-HR 2069-05-RT 2070-05-HR 2070-05-RT 2139-05-RT 2149P-05-RT 2179-05-RT
732-1166EA 80.4 EUR
732-1166 732-1181 732-1177 732-1175 732-1173 732-2391 732-2393 732-2394 732-2401 732-2403 732-2407
Barrier tips, low retention, ART™
Pipette Tips
These sterile barrier tips are designed to fit virtually any pipette and application. The self-sealing barrier provides 100% protection against aerosols and liquid contamination. Using advanced low-retention technology, the hydrophobic inner surface of these tips significantly reduces sample loss to adhesion and maximises sample recovery.
  • Free from RNase, DNase, DNA, ATP, bioburden, endotoxin, pyrogens and PCR inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks, ensuring safe and secure storage
  • Space-saving, stackable reload system is environmentally friendly, traceable and easy to use
Cdt: Tips are available in racks or in trays for rack reloading. Reload tips require an empty rack.