Imprimer
Overall, Pro Industry, FAS® button
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1003205412XL 1003205412XLSH 1003205413XLSH 100320541L 100320541L/SH 100320541L/T 100320541M 100320541M/SH 100320541M/T 100320541S 100320541S/SH 100320541S/T 100320541XL 100320541XL/SH 100320541XS
KWIN1003205412XLEA 140 EUR
KWIN1003205412XL KWIN1003205412XLSH KWIN1003205413XLSH KWIN100320541L KWIN100320541L/SH KWIN100320541L/T KWIN100320541M KWIN100320541M/SH KWIN100320541M/T KWIN100320541S KWIN100320541S/SH KWIN100320541S/T KWIN100320541XL KWIN100320541XL/SH KWIN100320541XS
Overall, Pro Industry, FAS® button
Tenue de travail
Durable overalls made of FAS® twill consisting of 100% cotton.
  • Concealed buttons at front
  • Two chest pockets with flap, one with pen pocket
  • Two front pockets; two back pockets, one with flap
  • Leg pocket over side seam
  • With inside folding-ruler pocket and three extra pockets
  • Leg pocket with flap, mobile phone pocket with flap, knee pockets
  • Adjustable waist and cuffs
Weight: 375 g/m²