Imprimer
Safety helmet, Endurance Plus, PW54
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PW54RBR PW54RER PW54WHR PW54YER
PRTWPW54RBREA 22.2 EUR
PRTWPW54RBR PRTWPW54RER PRTWPW54WHR PRTWPW54YER
Safety helmet, Endurance Plus, PW54
Casques Casques de sécurité
This fully certified ABS helmet features molten metal protection with a retractable visor.
  • Six-point self-sizing wheel ratchet suspension system
  • Can be worn in temperatures of −30 °C
Size: 520 to 620 mm
EN 397
Certifications: EN 397, EN166 1 B 9