Imprimer
Rain trousers, Sealtex® Classic, S451
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S451OGRL S451OGRXL S451BKRL S451BKRS S451BKRXL S451BKRM S451BKRXXL
PRTWS451OGRLEA 41.3 EUR
PRTWS451OGRL PRTWS451OGRXL PRTWS451BKRL PRTWS451BKRS PRTWS451BKRXL PRTWS451BKRM PRTWS451BKRXXL
Rain trousers, Sealtex® Classic, S451
Tenue de travail
Durable trousers made of stretchable Sealtex® Classic fabric consisting of 100% polyester tricot knit with PU coating. The fabric is tear-resistant, anti-fungal, oil-, fat-, diesel-, and grease-resistant, and can be easily wiped clean.
  • Stud adjustment at hem
  • High frequency welded waterproof seams for maximum water resistance
  • Elasticated waist
Weight: 200 g/m²