Imprimer
Shorts, Portwest Texo TX14
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TX14GRRXXL TX14GRRL TX14GRRM TX14GRRS TX14GRRXL TX14BKRL TX14BKRS TX14BKRM TX14BKRXL TX14BKRXXL TX14BKRXS TX14NARXL TX14NARM TX14NARS TX14NARXXL TX14NARL TX14NARXS
PRTWTX14GRRXXLEA 28.9 EUR
PRTWTX14GRRXXL PRTWTX14GRRL PRTWTX14GRRM PRTWTX14GRRS PRTWTX14GRRXL PRTWTX14BKRL PRTWTX14BKRS PRTWTX14BKRM PRTWTX14BKRXL PRTWTX14BKRXXL PRTWTX14BKRXS PRTWTX14NARXL PRTWTX14NARM PRTWTX14NARS PRTWTX14NARXXL PRTWTX14NARL PRTWTX14NARXS
Shorts, Portwest Texo TX14
Tenue de travail
These shorts are made of 60% cotton and 40% polyester to combine function and style. The soft handle cotton rich fabric is built to last. Cotton ensures it washes well at sixty degrees whilst the polyester gives a ruggedness essential for daily use.
  • Back elasticated waist
  • Versatile pockets for multiple storage space
  • Mobile phone pocket
  • Thigh pocket
Weight: 245 g/m²