Imprimer
Cloning cylinders, PYREX®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3166-10 3166-6 3166-8
CORN3166-10EA 85.4 EUR
CORN3166-10 CORN3166-6 CORN3166-8
Cloning cylinders, PYREX®
Éprouvettes Cylindres de clônage
Cloning cylinders, made of borosilicate glass code 7740. Clone a single cell or group of cells by surrounding them with this open ended cloning cylinders. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well.
  • Reusable
  • Non sterile
  • Cylinders are autoclavable or can be dry heat sterilised