Imprimer
Safety clogs, Steelite™ MICROFIBRE FW82, FW82
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FW82BKR36 FW82BKR35 FW82BKR39 FW82WHR37 FW82WHR48 FW82WHR47 FW82WHR46 FW82WHR45 FW82WHR44 FW82WHR43 FW82WHR42 FW82WHR41 FW82WHR40 FW82BKR37 FW82WHR38 FW82BKR38 FW82WHR36 FW82WHR35 FW82BKR48 FW82BKR47 FW82BKR46 FW82BKR45 FW82BKR44 FW82BKR43 FW82BKR42 FW82BKR41 FW82BKR40 FW82WHR39
PRTWFW82BKR36EA 50.5 EUR
PRTWFW82BKR36 PRTWFW82BKR35 PRTWFW82BKR39 114-4929 114-4940 114-4939 114-4938 114-4937 114-4936 114-4935 114-4934 114-4933 114-4932 PRTWFW82BKR37 114-4930 PRTWFW82BKR38 114-4928 114-4927 PRTWFW82BKR48 PRTWFW82BKR47 PRTWFW82BKR46 PRTWFW82BKR45 PRTWFW82BKR44 PRTWFW82BKR43 PRTWFW82BKR42 PRTWFW82BKR41 PRTWFW82BKR40 114-4931
Safety clogs, Steelite™ MICROFIBRE FW82, FW82
Chaussures et bottes
Anti-static lab clog manufactured from high quality, water resistant microfibre plus a single-density lightweight PU sole, resistant to oil.
  • Padded collars provide all day comfort
  • Detachable heel strap
  • Shock absorbent heel
  • Machine washable at 30 °C
  • Self-cleaning sole to prevent cross-contamination
  • Protective steel toecap (200 J)
For medical and food industry.
These shoes do not utilise animal product or biproducts.
Colour: Black or white
Certifications: EN ISO 20345:2011; SB, AE, WRU.