Imprimer
Safety cabinets with extinguisher for flammable products
Fournisseur: TRIONYX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AMS20 AMS100 AMS200 AMS10
TRIOAMS20EA 1100 EUR
TRIOAMS20 TRIOAMS100 TRIOAMS200 TRIOAMS10
Safety cabinets with extinguisher for flammable products
Mobilier Armoires de stockage
Safety cabinets with extinguisher for flammable products are equipped with a CO₂ extinguisher.
  • Large retention tank for AMS 100 and AMS 200 models
  • Automatic self-extinguishing in case of fire by thermic bulb
  • Sloping roof to avoid unauthorized storage
  • High and low vent Ø 100 mm with flame arresters
  • Self-closing double doors and key locking system
They are built in steel 15/10th and coated with epoxy paint beige RAL 1013 for AMS10 and AMS 20, yellow RAL 1023 for AMS100 and AMS 200.
These cabinet features with three or one adjustable perforated shelves respectively and bottom containment sump.
Certifications: Conforms with ISO 3864 and 92/58/CEE European directive.
Informations de commande: Available with additional perforated shelf for AMS100 and AMS200 models, spare extinguisher and supplied with 'Flammable' large warning labels.