Imprimer
Portable shower stations
Fournisseur: TRIONYX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DPA6 DPA9 CDPA6 CDPA9
118-0204EA 311 EUR
118-0204 118-0205 118-0200 118-0201
Portable shower stations
Matériel de premiers secours Douches
Portable shower stations with tested tank, made of E 243 and E 244 steel. Capacity 6 or 9 litres.
  • Spraying shower in polyamide and fibre glass
  • Green epoxy paint
  • Guidelines for usage printed on tank
Ideal for the chemical and pharmaceutical industry, agribusiness and automotive, chemistry laboratories, petrochemical and biochemistry laboratories, municipal workshops, garages and printing workshops, universities, high schools, colleges, hospitals, clinics, sewage treatment plants and pools.
Informations de livraison: Delivered with one CO₂ cartridge (40 or 60 g) installed. Replacement cartridges are available separately.
Attention: Delivered without liquid. Necessary to fill up with mineral water. Do not use in direct contact with skin or eyes.