Imprimer
Transport container with screw cap
Fournisseur: BOETTGER PAUL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
08.013.0000 08.013.0600 08.023.0600 08.033.0600 08.013.0400.04 08.023.0000 08.023.0400.04 08.033.0000 08.033.0400.04
BOET08.013.0000EA 116 EUR
BOET08.013.0000 BOET08.013.0600 BOET08.023.0600 BOET08.033.0600 BOET08.013.0400.04 BOET08.023.0000 BOET08.023.0400.04 BOET08.033.0000 BOET08.033.0400.04
Transport container with screw cap
Récipients de collecte d'échantillon Conteneurs à usage universel
Made of PP, translucent. Mailing containers for medical and biohazard samples.
  • Optimal protection from external elements through cylindrical design
  • Absorbent paper insert provides additional protection, easy to change, multiple-use very simple
  • Biohazard and recycling symbol clearly visible
Mailing containers Ø 30 mm fits into current mailboxes with 35 mm-wide slot, containers 44/114 mm suitable for mailing common culture mediums and containers 30/126 mm with cap suitable for Port-a-Cul.
Certifications: Corresponding to DIN EN 829 (with insert/print)En savoir plus