Imprimer
Sample tubes for stool, with screw cap
Fournisseur: BOETTGER PAUL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07.023.2007 07.043.2007 07.043.3107 07.033.2007 07.023.3107 07.033.3107
BOET07.023.2007EA 315 EUR
BOET07.023.2007 BOET07.043.2007 BOET07.043.3107 BOET07.033.2007 BOET07.023.3107 BOET07.033.3107
Sample tubes for stool, with screw cap
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
Clair tube with overlapping screw cap for optimal hygiene when opening, reliably sealed with long-lasting spoon for about bean-sized samples.
Certifications: Conforms to DIN EN 829 for the dispatch of stool samples.
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904