Imprimer
Luminescent/optical dissolved oxygen (LDO) electrodes, IntelliCAL™
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LDO10105 LDO10101 LDO10103 LDO10110 LDO10115 LDO10130
664-0208EA 1410 EUR
664-0208 664-0206 664-0207 664-0209 664-0210 664-0211
Luminescent/optical dissolved oxygen (LDO) electrodes, IntelliCAL™
Electrodes Dissolved Oxygen Electrodes
IntelliCAL™ LDO101 is a digital, luminescent dissolved oxygen (LDO) electrode which is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
  • Rugged digital probes can reliably test in the harshest environmental conditions: Stainless steel body, polymer sensor shroud, and reinforced, steel-sheathed cables protect the electrode for measurement anywhere in the field
  • No membranes to clean or replace, no electrolyte solution to replenish, and reduce calibration frequency
  • Ultimate traceability: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Alert the user when re-calibration is needed
  • Electrodes can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Ideal for a multi-user environment with multiple HQd series meters
The standard version is intended for laboratory use, the rugged version is intended for field use.