Imprimer
Disposable Post Mortem scalpels
Fournisseur: SWANN MORTON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2580 2581
SWAN2580EA 112 EUR
SWAN2580 SWAN2581
Disposable Post Mortem scalpels
Bistouris et pièces détachées Bistouris
Carbon blade. Non sterile, disposable histopathology and autopsy knives.
  • With easy to remove guard to protect the cutting edge: Users do not have to fix the blade to the handle
  • Available with either standard PM40 blade or PM40B bull nose blade fixed in a distinctive blue handle