Imprimer
Combined metal electrodes, Radiometer Analytical
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E31M002 E34M002 E31M003 E34M003
HACHE31M002EA 285 EUR
HACHE31M002 HACHE34M002 HACHE31M003 HACHE34M003
Combined metal electrodes, Radiometer Analytical
Électrodes Électrodes, autres
The Radiometer Analytical electrode range features single, double, and combined metal electrodes with platinum, silver, gold, antimony, mercury and glassy carbon sensing elements. This variety of designs can be used for redox measurements, redox titrations, potentiometric techniques, and imposed current potentiometric titration.
Informations de commande: The screw cap electrodes require the CL114 cable for use with Hach sensION+ benchtop and other BNC-compatible meters/titrators.