Imprimer
Scoops, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SWL028 SWL040
AZLOSWL028EA 42 EUR
AZLOSWL028 AZLOSWL040
Scoops, Azlon®
Pelles
Sturdy scoops made from natural rigid PE.
  • Lightweight
  • Good chemical resistance
  • Food safe