Imprimer
Standard opening screw thread vials
Fournisseur: THERMO NATIONAL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C4013-1 C4013-2 C4013-11
NSCAC4013-1EA 41.4 EUR
NSCAC4013-1 NSCAC4013-2 NSCAC4013-11
Standard opening screw thread vials
Fioles
Preferred for use with many carousel tray autosamplers. Glass vials are manufactured from superior quality borosilicate clear glass Type 1.
  • Re-usable sleeves
Microsampling and high recovery vials allow maximum sample extraction without need for separate inserts.