Imprimer
Dropping bottle, with tip and cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
211632 211620 211621
WHEA211632EA 311 EUR
WHEA211632 WHEA211620 WHEA211621
Dropping bottle, with tip and cap, WHEATON®
Flacons Flacons compte-gouttes
Dropping bottles are made of LDPE (low density polyethylene).
Dropping bottles with tips can be used to dispense a stream of liquid or a non-controlled drop. Dropping bottles, tips and caps are packaged separately.
The extended controlled dropper tip dispneses one drop at a time, with an average drop size of 44 µl ±6 µl. Radiation grade polypropylene screw caps and tips packaged separately. Dropping bottles, natural tips and white caps are packaged separately.