Imprimer
Industrial gloves, Titan 397
Fournisseur: MAPA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34397006 34397007 34397008 34397009 34397000
112-5580EA 69.6 EUR
112-5580 112-5581 112-5582 112-5583 112-5579
Industrial gloves, Titan 397
Gants
These nitrile gloves with textile support and smooth finish provide comfort and dexterity for common handling tasks.
  • Excellent comfort, due to the thickness of the cotton lining and the ventilated back
  • Freedom of movement: dexterity and flexibility, due to the close fit to the hand
  • Good abrasion resistance
Ideal for sanding, precision mechanical work, part assembly, handling oily mechanical parts, manufacturing small electrical components and for construction materials handling.
Certifications: EN 388:2016 (4111X); Cat. II