Imprimer
Industrial gloves, Ultranitril 472
Fournisseur: MAPA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34472260 34472266 34472247 34472268 34472269
MAPA34472260EA 30.3 EUR
MAPA34472260 MAPA34472266 MAPA34472247 MAPA34472268 MAPA34472269
Industrial gloves, Ultranitril 472
Gants
These nitrile gloves with chlorinated interior finish and pebbled exterior finish are suitable for use in the food industry, mechanical industry and in the construction industry (carpenters/joiners). They can be worn alone or as an outer-glove.
  • Fingertip precision for light chemical protection and food handling
  • Freedom of movement and excellent sensitivity, due to the gloves reduced thickness
  • Chlorinated finish for better protection of the products being handled
  • Suitable for all types of food
Certifications: EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved